(2-Bromophenyl)-(2,2-difluorocyclopropyl)methanol 95%
Catalog # APOSPC520848-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:263.079 g/mol
Storage Temperature:Ambient
CAS Number:1350637-25-0
Formula:C₁₀H₉BrF₂O
MDL Number:MFCD20492607