Order Entry
Italy
ContactUsLinkComponent
 
1-(Difluoromethoxy)-2,3-difluorobenzene 95%
Catalog # APOSPC520766-10G
Supplier:  Apollo Scientific
CAS Number:  
1-(Difluoromethoxy)-2,3-difluorobenzene 95%
Catalog # APOSPC520766-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :180.1 g/mol
Storage Temperature:Ambient
CAS Number:1404194-70-2
Formula:C₇H₄F₄O
MDL Number:MFCD22580881