
2-Fluoro-4-methyl-5-nitrophenyl trifluoromethanesulphonate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1980076-30-9
:
Formula:C₈H₅F₄NO₅S
MDL Number:MFCD28897190
:303.188 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together