Ethyl 1-[2-cyano-4-(difluoromEthyl)phenyl]piperidine-4-carboxylate
Catalog # APOSPC520319-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:308.323 g/mol
Storage Temperature:Ambient
Formula:C₁₆H₁₈F₂N₂O₂
MDL Number:MFCD30496633