5-(Difluoromethyl)-6-methoxy-1,3-benzodioxole
Catalog # APOSPC520308-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:202.155 g/mol
Storage Temperature:Ambient
CAS Number:2149598-37-6
Formula:C₉H₈F₂O₃
MDL Number:MFCD29767777