Order Entry
Italy
ContactUsLinkComponent
tert-Butyl 4-[4-(difluoromethyl)thiazol-2-yl]piperidine-1-carboxylate
tert-Butyl 4-[4-(difluoromethyl)thiazol-2-yl]piperidine-1-carboxylate
Catalog # APOSPC520270-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 4-[4-(difluoromethyl)thiazol-2-yl]piperidine-1-carboxylate
Catalog # APOSPC520270-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :318.383 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₂₀F₂N₂O₂S
MDL Number:MFCD29767751