Order Entry
Italy
ContactUsLinkComponent
2-(Difluoromethyl)-3-methoxy-thiophene
2-(Difluoromethyl)-3-methoxy-thiophene
Catalog # APOSPC520256-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Difluoromethyl)-3-methoxy-thiophene
Catalog # APOSPC520256-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :164.173 g/mol
Storage Temperature:Ambient
CAS Number:2149591-23-9
Formula:C₆H₆F₂OS
MDL Number:MFCD29767736