Order Entry
Italy
ContactUsLinkComponent
2-(Difluoromethyl)thieno[3,2-c]pyridine
2-(Difluoromethyl)thieno[3,2-c]pyridine
Catalog # APOSPC520254-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Difluoromethyl)thieno[3,2-c]pyridine
Catalog # APOSPC520254-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :185.194 g/mol
Storage Temperature:Ambient
Formula:C₈H₅F₂NS
MDL Number:MFCD29767734