Vai a Intestazione
Vai a Contenuti principale
Vai a Piè di pagina
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Difluoromethyl)-4-methyl-thiophene
2-(Difluoromethyl)-4-methyl-thiophene
2-(Difluoromethyl)-4-methyl-thiophene
APOSPC520252-1G
:
Apollo Scientific
:
2-(Difluoromethyl)-4-methyl-thiophene
APOSPC520252-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
148.174 g/mol
:
Ambient
:
C₆H₆F₂S
:
MFCD29767732