Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 5-(difluoromEthyl)thiophene-3-carboxylate
Ethyl 5-(difluoromEthyl)thiophene-3-carboxylate
Ethyl 5-(difluoromEthyl)thiophene-3-carboxylate
APOSPC520250-1G
:
Apollo Scientific
:
Ethyl 5-(difluoromEthyl)thiophene-3-carboxylate
APOSPC520250-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
206.21 g/mol
:
Ambient
:
189331-48-4
:
C₈H₈F₂O₂S
:
MFCD29767729