
2-Chloro-5-(difluoromethyl)thiophene
Catalog # APOSPC520248-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:168.592 g/mol
:GHS07
Storage Temperature:Refrigerator
Formula:C₅H₃ClF₂S
MDL Number:MFCD29767727