Order Entry
Italy
ContactUsLinkComponent
5-(Difluoromethyl)thiophene-2-carbonitrile
5-(Difluoromethyl)thiophene-2-carbonitrile
Catalog # APOSPC520235-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(Difluoromethyl)thiophene-2-carbonitrile
Catalog # APOSPC520235-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :159.157 g/mol
Storage Temperature:Ambient
CAS Number:2112657-85-7
Formula:C₆H₃F₂NS
MDL Number:MFCD29767715