Order Entry
Italy
ContactUsLinkComponent
2-(Difluoromethyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine hydrochloride
2-(Difluoromethyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine hydrochloride
Catalog # APOSPC520233-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Difluoromethyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine hydrochloride
Catalog # APOSPC520233-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :225.687 g/mol
Storage Temperature:Ambient
Formula:C₈H₉F₂NS·ClH
MDL Number:MFCD29767713