Order Entry
Italy
ContactUsLinkComponent
tert-Butyl 2-(difluoromethyl)-6,7-dihydro-4H-thieno[3,2-c]pyridine-5-carboxylate
tert-Butyl 2-(difluoromethyl)-6,7-dihydro-4H-thieno[3,2-c]pyridine-5-carboxylate
Catalog # APOSPC520232-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 2-(difluoromethyl)-6,7-dihydro-4H-thieno[3,2-c]pyridine-5-carboxylate
Catalog # APOSPC520232-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :289.341 g/mol
Storage Temperature:Ambient
Formula:C₁₃H₁₇F₂NO₂S
MDL Number:MFCD29767712