2-(2-Bromophenyl)-5-(difluoromethyl)thiophene
Catalog # APOSPC520201-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:289.139 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₇BrF₂S
MDL Number:MFCD29767682