2-(Difluoromethyl)-3-phenoxy-thiophene
Catalog # APOSPC520200-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:226.242 g/mol
Storage Temperature:Ambient
CAS Number:2149598-79-6
Formula:C₁₁H₈F₂OS
MDL Number:MFCD29767681