Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Difluoromethyl)-5-methylsulphonyl-thiophene
2-(Difluoromethyl)-5-methylsulphonyl-thiophene
2-(Difluoromethyl)-5-methylsulphonyl-thiophene
APOSPC520195-1G
:
Apollo Scientific
:
2-(Difluoromethyl)-5-methylsulphonyl-thiophene
APOSPC520195-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
212.237 g/mol
:
Ambient
:
C₆H₆F₂O₂S₂
:
MFCD29767676