7-(Difluoromethyl)-2,3-dimethyl-1H-indole
Catalog # APOSPC520119-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:195.209 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₁₁F₂N
MDL Number:MFCD29767602