Ethyl 3-(difluoromEthyl)-5-Ethyl-1H-indole-2-carboxylate
Catalog # APOSPC520108-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:267.271 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₅F₂NO₂
MDL Number:MFCD29767591