4-[3-(Difluoromethyl)-2-pyridyl]morpholine
Catalog # APOSPC520036-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:214.212 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₁₂F₂N₂O
MDL Number:MFCD29767520