Inserisci ordine
Italy
ContactUsLinkComponent
 
Ethyl 1-[3-(difluoromEthyl)-2-pyridyl]piperidine-4-carboxylate
Catalog # APOSPC520028-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 1-[3-(difluoromEthyl)-2-pyridyl]piperidine-4-carboxylate
Catalog # APOSPC520028-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Tipo di conf.:
    Bottle
  • Conf:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :284.302 g/mol
Storage Temperature:Ambiente
Formula:C₁₄H₁₈F₂N₂O₂
MDL Number:MFCD29767512