
2-Bromo-4-chloro-6-fluorobenzoic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1082040-61-6
:
Formula:C₇H₃BrClFO₂
MDL Number:MFCD11845467
:253.453 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together