3-Bromo-4-(difluoromethoxy)phenylacetic acid
Catalog # APOSPC510304-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:281.051 g/mol
Storage Temperature:Ambient
CAS Number:2006277-18-3
Formula:C₉H₇BrF₂O₃
MDL Number:MFCD29065908