Order Entry
Italy
ContactUsLinkComponent
2-Fluoro-6-(6-methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)benzaldehyde
2-Fluoro-6-(6-methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)benzaldehyde
Catalog # APOSPC510293-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Fluoro-6-(6-methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)benzaldehyde
Catalog # APOSPC510293-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :279.029 g/mol
Storage Temperature:Ambient
Formula:C₁₂H₁₁BFNO₅
MDL Number:MFCD29049232