5-Amino-6-fluoro-1,3-benzodioxole
Catalog # APOSPC510187-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:155.126 g/mol
Storage Temperature:Ambient
CAS Number:1366234-03-8
Formula:C₇H₆FNO₂
MDL Number:MFCD22069751