Order Entry
Italy
ContactUsLinkComponent
2-Propoxy-5-(trifluoromethyl)benzaldehyde
2-Propoxy-5-(trifluoromethyl)benzaldehyde
Catalog # APOSPC510040-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Propoxy-5-(trifluoromethyl)benzaldehyde
Catalog # APOSPC510040-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :232.199 g/mol
Storage Temperature:Ambient
CAS Number:472809-74-8
Formula:C₁₁H₁₁F₃O₂
MDL Number:MFCD06248747