Order Entry
Italy
ContactUsLinkComponent
7-(5-Bromo-2-fluorophenyl)-5,8-diazaspiro[3.4]octan-6-one
7-(5-Bromo-2-fluorophenyl)-5,8-diazaspiro[3.4]octan-6-one
Catalog # APOSPC510034-5G
Supplier:  Apollo Scientific
CAS Number:  
7-(5-Bromo-2-fluorophenyl)-5,8-diazaspiro[3.4]octan-6-one
Catalog # APOSPC510034-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :299.139 g/mol
Storage Temperature:Ambient
CAS Number:1272755-91-5
Formula:C₁₂H₁₂BrFN₂O
MDL Number:MFCD18651744