Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Ethyl 4,5-Difluoro-2-nitrobenzoate

Supplier:   Apollo Scientific
Ethyl 4,5-Difluoro-2-nitrobenzoate
Supplier:  Apollo Scientific
Certificates
Ethyl 4,5-Difluoro-2-nitrobenzoate

4 Options Available Below

Product Details & Documents
  • CAS Number:1214387-06-0
  • Formula:C₉H₇F₂NO₄
  • MDL Number:MFCD11042826
  • Molecular Weight:231.153 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 71,00 €
    71,00 €
    1 * 5 g
       71,00 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 354,00 €
    354,00 €
    1 * 25 g
       354,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 1 410,00 €
    1 410,00 €
    1 * 100 g
       1 410,00 €

    Specifications:
    New Product
    New Product
Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..