2-Deoxy-2,2-difluoro-D-glucose
Catalog # APOSPC50973-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Tipo di conf.:Bottle
- Conf:1 g
- Seq:3047
Specifications
About this item
:
:200.137 g/mol
Storage Temperature:Ambiente
CAS Number:34245-90-4
Formula:C₆H₁₀F₂O₅
MDL Number:MFCD30062869