Order Entry
Italy
ContactUsLinkComponent
1,2-Diethynyl-3-fluorobenzene
1,2-Diethynyl-3-fluorobenzene
Catalog # APOSPC50550-5G
Supplier:  Apollo Scientific
CAS Number:  
1,2-Diethynyl-3-fluorobenzene
Catalog # APOSPC50550-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :144.145 g/mol
Storage Temperature:Refrigerator
CAS Number:412041-38-4
Formula:C₁₀H₅F