Order Entry
Italy
ContactUsLinkComponent
2-Amino-5-bromo-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-5-bromo-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502867-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Amino-5-bromo-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502867-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :386.087 g/mol
Storage Temperature:Ambient
CAS Number:2244086-18-6
Formula:C₁₃H₆BrF₆NO
MDL Number:MFCD31630076