
3-Bromo-2-hydroxy-5-(trifluoromethoxy)benzaldehyde 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2149601-02-3
:
Formula:C₈H₄BrF₃O₃
MDL Number:MFCD30730320
:285.015 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together