Order Entry
Italy
ContactUsLinkComponent
Methyl 2-bromo-3,6-difluorobenzoate 97%
Methyl 2-bromo-3,6-difluorobenzoate 97%
Catalog # APOSPC501775-250MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-bromo-3,6-difluorobenzoate 97%
Catalog # APOSPC501775-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :251.025 g/mol
Storage Temperature:Ambient
CAS Number:1805523-44-7
Formula:C₈H₅BrF₂O₂
MDL Number:MFCD28061358