
4-Bromo-2-(trifluoromethyl)benzyl bromide 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:335013-18-8
:
Formula:C₈H₅Br₂F₃
MDL Number:MFCD13185414
:317.929 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together