
2,6-Difluoro-4-(trifluoromethyl)benzaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:134099-34-6
:
Formula:C₈H₃F₅O
MDL Number:MFCD12404959
:210.101 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together