
2-Chloro-4-fluoro-5-iodophenyl trifluoromethanesulphonate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1820607-92-8
:
Formula:C₇H₂ClF₄IO₃S
MDL Number:MFCD27937160
:404.505 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together