
4-Fluoro-2-hydroxy-5-(trifluoromethyl)benzaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1823917-95-8
:
Formula:C₈H₄F₄O₂
MDL Number:MFCD27664923
:208.11 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together