
2,4-Difluoro-5-(trifluoromethyl)benzaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:134099-31-3
:
Formula:C₈H₃F₅O
MDL Number:MFCD27664920
:210.101 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together