
4-Methyl-2-nitrophenyl trifluoromethanesulphonate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:195455-54-0
:
Formula:C₈H₆F₃NO₅S
MDL Number:MFCD25541871
:285.197 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together