
2-Methyl-3-nitrophenyl trifluoromethanesulphonate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1446016-49-4
:
Formula:C₈H₆F₃NO₅S
MDL Number:MFCD25541868
ADR:8,III
:285.197 g/mol
UN:3261
:GHS05
Storage Temperature:Ambiente
Frequently Bought Together