
6-Bromo-2-fluoro-3-nitrobenzotrifluoride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1440535-12-5
:
Formula:C₇H₂BrF₄NO₂
MDL Number:MFCD24850045
:287.994 g/mol
Storage Temperature:Ambiente
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together