
2-Bromo-6-chloro-4-fluorobenzoic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1695489-54-3
:
Formula:C₇H₃BrClFO₂
MDL Number:MFCD28733903
:253.453 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together