
3'-Bromo-2,2,2-trifluoro-5'-(trifluoromethyl)acetophenone 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1132701-00-8
:
Formula:C₉H₃BrF₆O
MDL Number:MFCD23098942
:321.014 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together