Ethyl 2-({[4-(trifluoromethyl)anilino]carbothioyl}amino)acetate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:321433-99-2
:
Formula:C₁₂H₁₃F₃N₂O₂S
MDL Number:MFCD00975552
:306.304 g/mol
Melting Pt:150…152 °C
Storage Temperature:Ambient
Frequently Bought Together