
3-(Dimethylsulphamoyl)-4-fluorobenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:381229-72-7
:
Formula:C₉H₁₀FNO₄S
MDL Number:MFCD09040991
:247.243 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together