
4-(Difluoromethyl)-2-(trifluoromethyl)benzaldehyde 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1453070-24-0
:
Formula:C₉H₅F₅O
MDL Number:MFCD31759999
:224.127 g/mol
:GHS05
Storage Temperature:Ambient
Frequently Bought Together