1-Bromo-4-iodo-2-(trifluoromethoxy)benzene 97%
Catalog # APOSPC48518-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:366.902 g/mol
Storage Temperature:Refrigerator
CAS Number:1187984-18-4
Formula:C₇H₃BrF₃IO
MDL Number:MFCD12761590