Order Entry
Italy
Orders LinkContactUsLinkComponent
2'-Methoxy-2,2,2,6'-tetrafluoroacetophenone 95%

2'-Methoxy-2,2,2,6'-tetrafluoroacetophenone 95%

Irritant
Supplier:   Apollo Scientific
2'-Methoxy-2,2,2,6'-tetrafluoroacetophenone 95%
Supplier:  Apollo Scientific
Certificates
2'-Methoxy-2,2,2,6'-tetrafluoroacetophenone 95%

2 Options Available Below

Product Details & Documents
Synonyms:  1-(2-Fluoro-6-methoxyphenyl)-2,2,2-trifluoroethan-1-one, 3-Fluoro-2-(trifluoroacetyl)anisole, 2,2,2-Trifluoro-1-(2-fluoro-6-methoxyphenyl)ethanone
  • CAS Number:1208078-28-7
  • Formula:C₉H₆F₄O₂
  • MDL Number:MFCD12922640
  • Molecular Weight:222.136 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 128,00 €
    128,00 €
    1 * 250 mg
       128,00 €

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 319,00 €
    319,00 €
    1 * 1 g
       319,00 €