
6-Hydroxy-2,2,3,3-tetrafluoro-1,4-benzodioxane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:103467-50-1
:
Formula:C₈H₄F₄O₃
MDL Number:MFCD00190145
:224.109 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together