


2,2-Difluoroethyl trifluoroacetate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7556-84-5
:
Formula:C₄H₃F₅O₂
MDL Number:MFCD19442057
:178.057 g/mol
Storage Temperature:Ambient
UN:2924
ADR:3+8,II
:GHS02
:GHS05
:GHS07
Frequently Bought Together